N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A127630-10MG
A127630-1MG
A127630-250MG
A127630-25G
A127630-25MG
A127630-50MG
A127630-10G
A127630-1G
A127630-5G
76797-744EA
6
USD
76797-744
76797-746
76797-748
76797-750
76797-752
76797-754
77599-852
77599-854
77599-856
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride 95%
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃₁H₄₅ClN₂O₅S |
MDL Number:
MFCD00914940 CAS Number: 141625-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...