(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: L-Arginine 4-nitroanilide dihydrochloride, H-Arg-pNA·2 HCl, L-Arginine p-nitroanilide dihydrochloride, H-Arg-pNA·2HCl

A127409-1G A127409-250MG A127409-25G A127409-5G
76797-736EA 125 USD
76797-736 76797-738 76797-740 76797-742
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride 97%
(S)-2-Amino-5-guanidino-N-(4-nitrophenyl)pentanamide dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₀Cl₂N₆O₃
MW: 367.231 g/mol
MDL Number: MFCD00058249
CAS Number: 40127-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR