Aloin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one, Barbaloin, 1,8-Dihydroxy-10-(beta-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone
A127344-100MG
A127344-1G
A127344-250MG
A127344-25G
A127344-5G
76797-718EA
5.31
USD
76797-718
76797-720
76797-722
76797-724
76797-726
Aloin 98%
Aloin
Precaución: for Research Use Only
Formula:
C₂₁H₂₂O₉ MW: 418.398 g/mol |
MDL Number:
MFCD00151160 CAS Number: 1415-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...