Dibutyl Fumarate 98%

Supplier: Ambeed
Warning

Synonyms: Dibutyl fumarate, Fumaric Acid Dibutyl Ester

A127002-100G A127002-25G A127002-500G
76797-704EA 14 USD
76797-704 76797-706 76797-708
Dibutyl Fumarate 98%
Dibutyl Fumarate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
MDL Number: MFCD00065141
CAS Number: 105-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR