2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol

A126824-100G A126824-25G A126824-1000G
76797-692EA 19.1 USD
76797-692 77313-098 77571-594
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole 97%
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O
MW: 323.44 g/mol
MDL Number: MFCD00013338
CAS Number: 3147-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR