D-Mannosamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Mannosamine HCl, (2S,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride, Mannosamine hydrochloride, 2-Amino-2-deoxy-D-mannose hydrochloride, D-Mannosamine HCl

A126596-10G A126596-1G A126596-25G A126596-5G A126596-100G A126596-250MG
76797-682EA 157.02 USD
76797-682 76797-684 76797-686 76797-688 77624-056 77624-058
D-Mannosamine hydrochloride 98%
D-Mannosamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₅
MW: 215.632 g/mol
MDL Number: MFCD00064557
CAS Number: 5505-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR