α-Isopropylphenylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: alpha-Isopropylphenylacetic Acid

A125729-1G A125729-5G A125729-25G
76797-662EA 29.71 USD
76797-662 77592-440 77599-818
α-Isopropylphenylacetic acid 95%
alpha-Isopropylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00021708
CAS Number: 3508-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR