1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acetyl-7(1H)-azaindole, 1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone

A125494-1G A125494-250MG A125494-5G
76797-648EA 33 USD
76797-648 76797-650 77878-919
1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone 98%
1-(1H-Pyrrolo[2,3-b]pyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD09743457
CAS Number: 83393-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR