4-Hydroxyquinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A125405-100G A125405-1G A125405-250MG A125405-25G A125405-5G
76797-638EA 787.19 USD
76797-638 76797-640 76797-642 76797-644 76797-646
4-Hydroxyquinoline-3-carboxylic acid 95%
4-Hydroxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.167 g/mol
MDL Number: MFCD00598887
CAS Number: 34785-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR