L-Threonine benzyl ester oxalate 98%

Supplier: Ambeed
Warning

Synonyms: H-Thr-OBzl·oxalate (1:1), (2S,3R)-Benzyl 2-amino-3-hydroxybutanoate oxalate

A125280-10G A125280-25G A125280-100G A125280-5G
76797-628EA 21 USD
76797-628 77566-410 77666-788 77666-790
L-Threonine benzyl ester oxalate 98%
L-Threonine benzyl ester oxalate

Precaución: For research use only.

Formula: C₁₃H₁₇NO₇
MDL Number: MFCD02259509
CAS Number: 201274-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR