6-(4-(1H-Imidazol-1-yl)phenyl)-4,5-dihydropyridazin-3(2H)-one 97%

Supplier: Ambeed
Warning

A125170-1G A125170-250MG
76797-624EA 36 USD
76797-624 76797-626
6-(4-(1H-Imidazol-1-yl)phenyl)-4,5-dihydropyridazin-3(2H)-one 97%
6-(4-(1H-Imidazol-1-yl)phenyl)-4,5-dihydropyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₄O
MDL Number: MFCD00867109
CAS Number: 84243-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR