1-Benzoylpiperazine 97%

Supplier: Ambeed
Warning

A124671-10G A124671-1G A124671-25G A124671-5G A124671-100G
76797-604EA 42 USD
76797-604 76797-606 76797-608 76797-610 77666-784
1-Benzoylpiperazine 97%
1-Benzoylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD00810192
CAS Number: 13754-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR