Methyl benzo[b]thiophene-2-carboxylate 97%

Supplier: Ambeed
Warning

A124578-1G A124578-250MG A124578-25G A124578-5G
76797-600EA 28.65 USD
76797-600 76797-602 77566-386 77666-782
Methyl benzo[b]thiophene-2-carboxylate 97%
Methyl benzo[b]thiophene-2-carboxylate

Precaución: For research use only.

Formula: C₁₀H₈O₂S
MW: 192.24 g/mol
MDL Number: MFCD00067791
CAS Number: 22913-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR