2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Danger

A124254-250MG A124254-5G A124254-1G
76797-594EA 21 USD
76797-594 76797-596 77666-774
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 97%
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₇H₈F₃N₃
MDL Number: MFCD09834983
CAS Number: 126069-70-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR