Dichlorodi(cyclopenta-1,3-dien-1-yl)platinate(II) 98%

Supplier: Ambeed
Warning

A123661-100MG A123661-1G A123661-250MG
76797-530EA 61.54 USD
76797-530 76797-532 76797-534
Dichlorodi(cyclopenta-1,3-dien-1-yl)platinate(II) 98%
Dichlorodi(cyclopenta-1,3-dien-1-yl)platinate(II)

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂Pt
MW: 396.17 g/mol
MDL Number: MFCD00269838
CAS Number: 12083-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR