1-Boc-piperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid

A123637-100G A123637-10G A123637-25G A123637-5G
76797-520EA 22 USD
76797-520 76797-522 76797-524 76797-526
1-Boc-piperidine-3-carboxylic acid 95%
1-Boc-piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
MDL Number: MFCD00673775
CAS Number: 84358-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR