L(+)-Histidine monohydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (2S)-(+)-Histidine monohydrochloride, L-3-Imidazol-4-ylalanine monohydrochloride, (S)-(+)-Histidine monohydrochloride, His HCl, (S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid hydrochloride, H-His-OH·HCl

A123193-100G A123193-10G A123193-25G A123193-500G
76797-512EA 20 USD
76797-512 76797-514 76797-516 76797-518
L(+)-Histidine monohydrochloride 97%
L(+)-Histidine monohydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃O₂
MW: 191.62
MDL Number: MFCD00064556
CAS Number: 645-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR