Eicosanedioic acid 98%

Supplier: Ambeed
Warning

A123107-1G A123107-250MG A123107-25G A123107-5G
76797-498EA 7 USD
76797-498 76797-500 76797-502 76797-504
Eicosanedioic acid 98%
Eicosanedioic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₈O₄
MDL Number: MFCD00059627
CAS Number: 2424-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR