2-Quinolinecarbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: Quinoline-2-carboxaldehyde, Quinoline-2-carbaldehyde, 2-Quinolinecarboxaldehyde

A123039-100G A123039-10G A123039-1G A123039-250MG A123039-25G A123039-5G
76797-486EA 409.51 USD
76797-486 76797-488 76797-490 76797-492 76797-494 76797-496
2-Quinolinecarbaldehyde 97%
2-Quinolinecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO
MW: 157.17 g/mol
MDL Number: MFCD00075032
CAS Number: 5470-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR