2-Bromo-1-(3-methylisoxazol-5-yl)ethanone 97%

Supplier: Ambeed
Danger

A122622-100MG A122622-250MG
76797-456EA 116.7 USD
76797-456 76797-458
2-Bromo-1-(3-methylisoxazol-5-yl)ethanone 97%
2-Bromo-1-(3-methylisoxazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
MW: 204.02 g/mol
MDL Number: MFCD11046397
CAS Number: 104777-32-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR