(R)-Methyl 3-((tert-butyldimethylsilyl)oxy)-5-oxo-6-(triphenylphosphoranylidene)hexanoate 97%

Supplier: Ambeed
Warning

A122597-1G A122597-5G A122597-10G
76797-450EA 14 USD
76797-450 76797-452 76797-454 77878-903
(R)-Methyl 3-((tert-butyldimethylsilyl)oxy)-5-oxo-6-(triphenylphosphoranylidene)hexanoate 97%
(R)-Methyl 3-((tert-butyldimethylsilyl)oxy)-5-oxo-6-(triphenylphosphoranylidene)hexanoate

Precaución: for Research Use Only

Formula: C₃₁H₃₉O₄PSi
MDL Number: MFCD09753068
CAS Number: 147118-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR