Isobutyl-3,5-diamino-4-chlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Isobutyl 3,5-diamino-4-chlorobenzoate

A122184-100G A122184-1G A122184-25G A122184-5G
76797-438EA 101.85 USD
76797-438 76797-440 76797-442 76797-444
Isobutyl-3,5-diamino-4-chlorobenzoate 98%
Isobutyl-3,5-diamino-4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O₂
MW: 242.71 g/mol
MDL Number: MFCD00272560
CAS Number: 32961-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR