Ethyl-4-bromobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate

A121373-100G A121373-10G A121373-25G A121373-500G A121373-5G
76797-400EA 43.5 USD
76797-400 76797-402 76797-404 76797-406 76797-408
Ethyl-4-bromobenzoate 98%
Ethyl-4-bromobenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00016329
CAS Number: 5798-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR