1-(Phenylsulfonyl)pyrrole 97%

Supplier: Ambeed
Warning

A121365-100G A121365-25G A121365-500G A121365-5G
76797-392EA 87 USD
76797-392 76797-394 76797-396 76797-398
1-(Phenylsulfonyl)pyrrole 97%
1-(Phenylsulfonyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂S
MDL Number: MFCD00067739
CAS Number: 16851-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR