H-9 HCl 97%

Supplier: Ambeed
Warning

A120903-1G A120903-250MG A120903-5G
76797-332EA 41 USD
76797-332 76797-334 76797-336
H-9 HCl 97%
H-9 HCl

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O₂S·HCl
MDL Number: MFCD00083242
CAS Number: 116970-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR