Sodium glutamate 95%

Supplier: Ambeed
Warning

Synonyms: Sodium (S)-2-amino-4-carboxybutanoate, MSG, Monosodium glutamate, L-Glutamic acid monosodium salt, H-Gly-OH(Sodium)

A120841-1000G A120841-100G A120841-500G
76797-320EA 31 USD
76797-320 76797-322 76797-324
Sodium glutamate 95%
Sodium glutamate

Precaución: for Research Use Only

Formula: C₅H₈NNaO₄
MDL Number: MFCD00013074
CAS Number: 142-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR