Geranyl tiglate 98+%

Supplier: Ambeed
Warning

Synonyms: Tiglic acid Geranyl Ester, (E)-(E)-3,7-Dimethylocta-2,6-dien-1-yl 2-methylbut-2-enoate

A120834-100G A120834-25G
76797-316EA 53.05 USD
76797-316 76797-318
Geranyl tiglate 98+%
Geranyl tiglate

Precaución: for Research Use Only

Formula: C₁₅H₂₄O₂
MW: 236.36 g/mol
MDL Number: MFCD00016622
CAS Number: 7785-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR