(5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol (S)-5-oxopyrrolidine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Metadoxine

A120689-100G A120689-10G A120689-25G A120689-5G
76797-308EA 143.23 USD
76797-308 76797-310 76797-312 76797-314
(5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol (S)-5-oxopyrrolidine-2-carboxylate 97%
(5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol (S)-5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₆
MW: 298.29 g/mol
MDL Number: MFCD08704232
CAS Number: 74536-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR