Ethyl 3-amino-3-(pyridin-3-yl)propanoate dihydrochloride 98%

Supplier: Ambeed
Warning

A120338-100MG A120338-250MG
76797-304EA 27.59 USD
76797-304 76797-306
Ethyl 3-amino-3-(pyridin-3-yl)propanoate dihydrochloride 98%
Ethyl 3-amino-3-(pyridin-3-yl)propanoate dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆Cl₂N₂O₂
MW: 267.15 g/mol
MDL Number: MFCD11053562
CAS Number: 149498-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR