Light green SF yellowish 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30
A118984-1G
A118984-5G
76797-256EA
14
USD
76797-256
76797-258
Light green SF yellowish 97%
Light green SF yellowish
Precaución: for Research Use Only
Formula:
C₃₇H₃₄N₂Na₂O₉S₃ MW: 792.86 g/mol |
MDL Number:
MFCD00012121 CAS Number: 5141-20-8 Merck Index: 13,05506 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...