3-Methylquinoline 97%

Supplier: Ambeed
Warning

A118498-100G A118498-1G A118498-5G A118498-10G A118498-25G
76797-232EA 266.29 USD
76797-232 76797-234 76797-236 77571-100 77666-698
3-Methylquinoline 97%
3-Methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉N
MW: 143.19 g/mol
MDL Number: MFCD00014661
CAS Number: 612-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR