2,3-Dihydro-1H-carbazol-4(9H)-one 97%

Supplier: Ambeed
Warning

Synonyms: Ondansetron Impurity I, 1,2,3,4-Tetrahydro-4-oxocarbazole, 1,2,3,4-Tetrahydrocarbazol-4-one

A118328-100G A118328-10G A118328-25G A118328-5G
76797-222EA 49.87 USD
76797-222 76797-224 76797-226 76797-228
2,3-Dihydro-1H-carbazol-4(9H)-one 97%
2,3-Dihydro-1H-carbazol-4(9H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.2218 g/mol
MDL Number: MFCD00173749
CAS Number: 15128-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR