4-Hydroxy-7-methoxyquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A118044-1G A118044-5G A118044-250MG
76797-212EA 12 USD
76797-212 76797-214 77878-883
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid 97%
4-Hydroxy-7-methoxyquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MDL Number: MFCD00024004
CAS Number: 28027-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR