1-Boc-2-ethylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-ethylpiperazine-1-carboxylate

A117967-100MG A117967-1G A117967-250MG A117967-5G
76797-196EA 22.28 USD
76797-196 76797-198 76797-200 76797-202
1-Boc-2-ethylpiperazine 97%
1-Boc-2-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
MDL Number: MFCD03265490
CAS Number: 393781-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR