Ethyl 3-(3,4-dihydroxyphenyl)acrylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl Caffeate
A117016-100G
A117016-100MG
A117016-10G
A117016-1G
A117016-5G
A117016-25G
A117016-50MG
76797-108EA
515.6
USD
76797-108
76797-110
76797-112
76797-114
76797-116
77599-690
77599-692
Ethyl 3-(3,4-dihydroxyphenyl)acrylate 98%
Ethyl 3-(3,4-dihydroxyphenyl)acrylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₄ MW: 208.21 g/mol |
MDL Number:
MFCD00045754 CAS Number: 102-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...