Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Diclofenac Diethylamine

A115940-25G A115940-500G A115940-50MG A115940-5G A115940-100G
76797-056EA 11.67 USD
76797-056 76797-058 76797-060 76797-062 77599-670
Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate 98%
Diethylamine 2-(2-((2,6-dichlorophenyl)amino)phenyl)acetate

Precaución: for Research Use Only

Formula: C₁₈H₂₂Cl₂N₂O₂
MW: 369.28 g/mol
MDL Number: MFCD01862249
CAS Number: 78213-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR