3-(Indol-3-yl)propionic acid 97%

Supplier: Ambeed
Warning

A115498-1000G A115498-100G A115498-10G A115498-25G A115498-500G A115498-5G
76797-044EA 187 USD
76797-044 76797-046 76797-048 76797-050 76797-052 76797-054
3-(Indol-3-yl)propionic acid 97%
3-(Indol-3-yl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD00005660
CAS Number: 830-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR