Pyrrolidin-3-ylmethanol hydrochloride 98%

Supplier: Ambeed
Warning

A115240-1G A115240-250MG A115240-5G A115240-25G
76797-038EA 24 USD
76797-038 76797-040 76797-042 77878-866
Pyrrolidin-3-ylmethanol hydrochloride 98%
Pyrrolidin-3-ylmethanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
MDL Number: MFCD09800469
CAS Number: 644971-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR