5-Fluoro-DL-tryptophan 95%

Supplier: Ambeed
Warning

A114530-100MG A114530-1G A114530-250MG A114530-5G
76797-022EA 32 USD
76797-022 76797-024 76797-026 77666-664
5-Fluoro-DL-tryptophan 95%
5-Fluoro-DL-tryptophan

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂O₂
MDL Number: MFCD00005649
CAS Number: 154-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR