Dibenzyl phosphate 98%

Supplier: Ambeed
Warning

A114513-10G A114513-1G A114513-25G A114513-5G A114513-100G A114513-500G
76797-014EA 10 USD
76797-014 76797-016 76797-018 76797-020 77570-834 77878-860
Dibenzyl phosphate 98%
Dibenzyl phosphate

Precaución: for Research Use Only

Formula: C₁₄H₁₅O₄P
MDL Number: MFCD00004775
CAS Number: 1623-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR