3,4-Dimethyl-1,2-cyclopentanedione 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethylcyclopentane-1,2-dione, 3,4-Dimethyl-1,2-cyclopentadione

A114376-100G A114376-25G A114376-5G
76797-008EA 319.34 USD
76797-008 76797-010 76797-012
3,4-Dimethyl-1,2-cyclopentanedione 97%
3,4-Dimethyl-1,2-cyclopentanedione

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD00143063
CAS Number: 13494-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR