(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride 97%

Supplier: Ambeed
Danger

A114018-100MG A114018-250MG
76796-980EA 55 USD
76796-980 76796-982
(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride 97%
(S)-3-Amino-1-chloro-5-methylhexan-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO·HCl
MDL Number: MFCD00238011
CAS Number: 54518-92-2
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR