1-Benzyl-3-aminopyrrolidine 96%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-1-benzylpyrrolidine, 1-Benzylpyrrolidin-3-amine

A113550-10G A113550-1G A113550-25G A113550-5G
76796-960EA 88.06 USD
76796-960 76796-962 76796-964 76796-966
1-Benzyl-3-aminopyrrolidine 96%
1-Benzyl-3-aminopyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00059036
CAS Number: 18471-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR