(±)-Dimethyl acetylsuccinate 96%

Supplier: Ambeed
Warning

Synonyms: Dimethyl acetylsuccinate

A113066-1000G A113066-100G A113066-25G A113066-500G
76796-952EA 146 USD
76796-952 76796-954 76796-956 76796-958
(±)-Dimethyl acetylsuccinate 96%
(±)-Dimethyl acetylsuccinate

Precaución: for Research Use Only

Formula: C₈H₁₂O₅
MDL Number: MFCD00008448
CAS Number: 10420-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR