Isopropyl laurate 98%

Supplier: Ambeed
Warning

Synonyms: Propan-2-yl dodecanoate, Isopropyl dodecanoate

A111861-5G A111861-100G A111861-25G
76796-900EA 5.31 USD
76796-900 76796-902 77666-630 77666-632
Isopropyl laurate 98%
Isopropyl laurate

Precaución: for Research Use Only

Formula: C₁₅H₃₀O₂
MW: 242.398 g/mol
MDL Number: MFCD00451146
CAS Number: 10233-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR