Congo red indicator

Supplier: Ambeed
Warning

Synonyms: CI 22120

A111810-100G A111810-25G A111810-5G
76796-894EA 20.16 USD
76796-894 76796-896 76796-898
Congo red indicator
Congo red

Precaución: for Research Use Only

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
MDL Number: MFCD00004028
CAS Number: 573-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR