(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid 98%

Supplier: Ambeed
Danger

Synonyms: (2-Amino-1,3-thiazol-4-yl)[(2-tert-butoxy-2-oxoethoxy)imino]acetic acid

A111140-10G A111140-1G A111140-250MG A111140-5G
76796-868EA 18.04 USD
76796-868 76796-870 76796-872 76796-874
(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid 98%
(Z)-2-(2-Aminothiazol-4-yl)-2-(tert-Butoxycarbonylmethoxyimino)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₅S
MW: 301.32 g/mol
MDL Number: MFCD06411064
CAS Number: 74440-02-1
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR