Dicyclopropyl ketone 98%

Supplier: Ambeed
Danger

Synonyms: Dicyclopropylketone, Dicyclopropylmethanone

A110848-25G A110848-5G A110848-100G
76796-830EA 27.59 USD
76796-830 76796-832 77592-186
Dicyclopropyl ketone 98%
Dicyclopropyl ketone

Precaución: for Research Use Only

Formula: C₇H₁₀O
MW: 110.16 g/mol
MDL Number: MFCD00001276
CAS Number: 1121-37-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR