Ethyl-3-hydroxybenzoate 98%

Supplier: Ambeed
Warning

A110678-25G A110678-100G A110678-10G A110678-5G A110678-1000G A110678-500G
76796-806EA 14 USD
76796-806 77599-594 77599-596 77599-598 77878-836 77878-837
Ethyl-3-hydroxybenzoate 98%
Ethyl-3-hydroxybenzoate
Formula: C₉H₁₀O₃
MDL Number: MFCD00002296
CAS Number: 7781-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR