(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-(3,5-Bis(trifluoromethyl)phenyl)ethanamine

A110217-1G A110217-250MG
76796-772EA 321.46 USD
76796-772 76796-774
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine 97%
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethylamine

Precaución: for Research Use Only

Formula: C₁₀H₉F₆N
MW: 257.176 g/mol
MDL Number: MFCD03093013
CAS Number: 127733-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR